Cut resistant gloves, HyFlex® 74-718
Supplier: Ansell
Certificates
About this item
These knitted gloves are made from stainless steel, spandex, glass fiber and Dyneema® yarn and provide reliable cut protection.
- Zonal plating along with Dyneema® Diamond Technology enhances protection in high-risk areas of the hand
- Incorporates Tuff Cuff™ II technology to retain ergonomic shape during use and after laundering
- KVSD (Knitted Variable Stitch Design) technology to reduce hand fatigue
- Advanced fibres to help control moisture and hand temperature in cold and warm environments
Ideal for slicing and dicing, filleting fish and food preparation, processing and handling.
Certifications: EN 388:2016 (4X4XE); Cat. II
Food contact approved
Food contact approved
Specifications
- Catalog No:
- 112-4627
- 112-4626
- 112-4624
- 112-4628
- 112-4625
- Model:
- 74-718
- 74-718
- 74-718
- 74-718
- 74-718
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Gauge:
- 10
- 10
- 10
- 10
- 10
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X4XE)
- EN 388:2016 (4X4XE)
- EN 388:2016 (4X4XE)
- EN 388:2016 (4X4XE)
- EN 388:2016 (4X4XE)
- Food contact approved:
- +
- +
- +
- +
- +