ContactUsLinkComponent
Buffer solution for water testing
Buffer solution for water testing
  HACH1430598
 :  Hach
 :  
undefined
Buffer solution for water testing
  HACH1430598
 :  Hach
 :  
  • Pk:
    25
  • Description:
    Magnesium chloride/potassium phosphate buffer solution, 99 ml
Buffered dilution water required for testing recreational, drinking and wastewater.