Demineraliser bottle
Catalog # HACH1429900
Supplier: Hach
Specifications
- Capacity:177 ml
- Description:Demineraliser bottle
- Pk:1
Specifications
About this item
This narrow mouth polyethylene demineraliser bottle is used to deionize small volumes of water.
- Flip spout cap
- Contain monobed resin inside the bottle