Order Entry
Germany
ContactUsLinkComponent
Single element standard for ICP, gold
Single element standard for ICP, gold
Catalog # HEWL8500-7000
Supplier:  Agilent
CAS Number:  
undefined
Single element standard for ICP, gold
Catalog # HEWL8500-7000
Supplier:  Agilent
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2-5% HNO₃
  • Pk:
    1
  • Element:
    Gold
  • Symbol:
    Au

Specifications

About this item