ContactUsLinkComponent
Escape hoods, Parat® 4700
Escape hoods, Parat® 4700
Catalog # 111-2170
Supplier:  Dräger
undefined
Escape hoods, Parat® 4700
Catalog # 111-2170
Supplier:  Dräger
Specifications
  • Description:
    Escape hood, Parat® 4730, with ABEK P3 filter, Hard Case
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This combined fire and industrial escape hood was developed with users, placing the focus on the fastest possible escape. Optimised operation and wearing comfort, a robust housing and a tested ABEK P3 filter ensure protection from toxic industrial gases, vapours and particles for at least 15 minutes.
  • Hood in signal colour for high visibility
  • Automatically adjusting internal head harness: No additional tighting of the hood is required
  • Flexible textile neck seal
  • Exhalation valve directly leads the exhaled air out of the half mask
  • Wide field of vision thanks to a specially formed visor with anti-fog coating
  • Filter is automatically released from the packaging when opened
  • Mask fits a wide variety of face sizes
When opening the packaging, the filter plug is automatically released from the filter. The filter is then deployed into operational position and the hood can be immediately donned. Thanks to the self-adjusting internal head harness, no additional adjustment is required. Users only have to open the packaging, remove and don the hood – and leave the danger zone.
Total service life: 16 years (provided the filter is exchanged after eight years)
Certifications: DIN 58647-7 (filter additionally tested in accordance with EN 14387:2004)
Packaging: Two packaging types are available: The Hard Case provides splash water protection (IP 54) – the Soft Pack provides dust protection (IP 5). Both packaging types have a viewing window to check the filter expiration date and the condition of the device. The security seal on the packaging shows if the unit has been opened.