QBT™ Fatty acid uptake assay kit
Supplier: Molecular Devices
Certificates
About this item
QBT™ Fatty Acid Uptake Assay Kit is a homogeneous assay amenable to high-throughput screening. The kits deliver pre-optimized, fluorescence-based formulations to expedite assay development and screening of fatty acid transporters.
- One-step, homogeneous assay format enables high-throughput screening
- Fluorescent assay with proprietary quench technology allows users to monitor real-time uptake kinetics in live cells
- Replaces cumbersome and time-consuming fluorescent and radioactive assay protocols
- Elimination of radioactive compounds reduces disposal costs and eliminates safety risks associated with radioactive assays