ContactUsLinkComponent
Protein ladders (pre-stained), ScanLater™
Protein ladders (pre-stained), ScanLater™
Catalog # MLDVR8220
Supplier:  Molecular Devices
CAS Number:  
undefined
Protein ladders (pre-stained), ScanLater™
Catalog # MLDVR8220
Supplier:  Molecular Devices
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    ScanLater™ Western Blot Protein Ladder (prestained)
Specifications
About this item
The ScanLater™ Western blot protein ladder is a mixture of seven biotinylated recombinant proteins at 10, 20, 40, 60, 80, 100, and 140 kD.
  • 3 pre-stained marker proteins at 18, 31, and 70 kD
  • Ready to use, for 30 mini gels with recommended protocol