Calibration standard, single element, uranium for AAS and ICP-OES, pure grade
PERCN9303844
:
- Konzentration:1000 mg/L
- Matrix:2% HNO₃
- VE:125 mL
- Element:Uran
- Symbol:U
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.