Order Entry
Germany
ContactUsLinkComponent
Calibration standard, single element, carbon for AAS and ICP-OES, pure grade
Calibration standard, single element, carbon for AAS and ICP-OES, pure grade
Catalog # PERCN9303762
Supplier:  PerkinElmer
CAS Number:  
Calibration standard, single element, carbon for AAS and ICP-OES, pure grade
Catalog # PERCN9303762
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    125 mL
  • Element:
    Carbon
  • Symbol:
    C

Specifications

About this item

Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.

Caution: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.