Order Entry
Germany
ContactUsLinkComponent
Urine collecting system, Monovette®
Urine collecting system, Monovette®
Catalog # SARS10.252.020
Supplier:  SARSTEDT
undefined
Urine collecting system, Monovette®
Catalog # SARS10.252.020
Supplier:  SARSTEDT

Specifications

  • Ø:
    15 mm
  • Capacity:
    10 ml
  • Description:
    Urine collecting system, Urine Monovette®
  • Length:
    102 mm
  • Pk:
    500

Specifications

About this item

Urine collection system specifically designed for transport and storage of urine.