Order Entry
Germany
ContactUsLinkComponent
Test kits, photometric, COD, LCK cuvette tests
Test kits, photometric, COD, LCK cuvette tests
Catalog # HACHLCK214
Supplier:  Hach
CAS Number:  
undefined
Test kits, photometric, COD, LCK cuvette tests
Catalog # HACHLCK214
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Dichromate
  • Pk:
    25 Tests
  • Description:
    COD Cuvette Test, mercury-free
  • Seq:
    04
  • Type:
    LCK214
  • For unit:
    DR6000, DR3900, DR1900
  • Range:
    100 - 1000 mg/l

Specifications

About this item

Cuvette tests for water testing applications.

  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding

Storage conditions: 15 to 25 °C (protect from light)

Certifications: ISO 6060-1989, DIN 38409-H41
Accessories information: Cuvette tests are officially approved for many legally required consent limits with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.