Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%
Certificates
undefined
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%

Specification Test Results

  • Appearance:
    White to yellow to tan Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    114 to 118 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Related Information