1,2-Benzisoxazol-3-amine 97%
ACRO451710050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Off-white to brown Solid
- Infrared spectrum:Conforms
- Melting point:107 to 112 °C
- HPLC:≥96.0 %
:
:134.1400 g/mol
:110…112 °C
:GHS07
:Ambient
:36216-80-5
:C₇H₆N₂O
:MFCD03407353