
2-Bromo-5-nitrothiophene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13195-50-1
:
Formula:C₄H₂BrNO₂S
MDL Number:MFCD00022493
:208.04 g/mol
Boiling Pt:235…237 °C (1013 hPa)
Melting Pt:44…48 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to tan Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:43 to 45 °C
- GC:≥96.0 %