Order Entry
Germany
ContactUsLinkComponent
 
 
2-Ethyl-3-hydroxy-4-pyrone 98%
 
undefined
 
2-Ethyl-3-hydroxy-4-pyrone 98%

 

  • Appearance:
    White to cream Crystalline powder or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    88 to 94 °C
  • HPLC:
    ≥97.5 %