Order Entry
Germany
ContactUsLinkComponent
 
 
2-Iminothiolane hydrochloride (Traut's Reagent) 98%
 
undefined
 
2-Iminothiolane hydrochloride (Traut's Reagent) 98%

 

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    193 to 198 °C
  • Titration Argentometric:
    ≥97.5 %

 

 

Frequently Bought Together