Diethyl ether, extra dry 99.5% stabilised, AcroSeal®
Glass bottle, 4× 25 ml, 100 mL
Supplier: Thermo Fisher Scientific
Diethyl ether, extra dry 99.5% stabilised, AcroSeal®
Catalog # 448421000.
Supplier: Thermo Fisher Scientific
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Package of 4x25ML bottles
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Boiling Pt:34.6 °C (760 mmHg)
- Melting Pt:–116 °C
- Density:0.714 g/cm³
- Flash Pt:–45 °C
- Storage Temperature:Ambient
- UN:1155
- ADR:3,I
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.005 % (Coulometric)
- GC:≥99.4 %
- Stabilizer:1.5 to 10 ppm BHT
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0002 meq/g
- Filt.over 0.2 micron PFTE:Filtered
- Peroxides:≤0.002 % (as H2O2)
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Pk Info:4× 25 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



