Acetonitrile, extra dry 99.9%, AcroSeal®
Catalog # 448391000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2320
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Pk Info:4× 25 ml
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.001 % (Coulometric)
- GC:≥99.85 %
- Residue after evaporation:≤0.0005 %
- Acidity (CH3COOH):≤0.002 %
Specification Test Results
About this item
:
:41.05 g/mol
Boiling Pt:81…82 °C (760 mmHg)
Melting Pt:–46 °C
Density:0.781 g/cm³
Flash Pt:2 °C
UN:1648
ADR:3,II
:GHS07
:GHS02
Storage Temperature:Ambient
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878