ContactUsLinkComponent
Potassium permanganate ACS low in mercury
Irritant
Environment
Oxidizing
Corrosive
Health
Potassium permanganate ACS low in mercury
Certificates
undefined
Irritant
Environment
Oxidizing
Corrosive
Health
Potassium permanganate ACS low in mercury
Specification Test Results
  • Appearance:
    Dark green to brown-black Crystalline powder
  • Titration with KMnO4:
    ≥99.0 %
  • Chloride and Chlorate (as Cl):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.02 %
  • Mercury (Hg):
    ≤0.05 ppm
  • Insoluble matter:
    ≤0.2 %