Ammonium acetate 98% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
:77.08 g/mol
Melting Pt:114 °C
Density:1,17 g/cm³ (20 °C)
Flash Pt:136 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Solubility:(1 M in water) Clear colorless
- UV:(1 M in water)
- pH:6.5 to 7.3 (5 % in water)
- Titration with HClO4:≥97.5 %
- Heavy metals (as Pb):≤2 ppm