Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rhodamine WT
Rhodamine WT 20% in water
Rhodamine WT 20% in water
:
Thermo Fisher Scientific
Rhodamine WT 20% in water
:
Thermo Fisher Scientific
:
37299-86-8
:
:
C₂₉H₂₉ClN₂Na₂O₅
:
MFCD01728973
:
1.16 g/cm³
:
566.99 g/mol
:
Ambient
Appearance:
Dark red Liquid
UV Lambda max:
553 to 557 nm
Strength:
19.5 to 21 % (dye content)
ACRO44697