
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:193140-71-5
:
Formula:C₅H₉ClF₃NO₂
MDL Number:MFCD14525479
:207.58 g/mol
Melting Pt:218…220 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Powder or solid
- Infrared spectrum:Conforms
- Titration Argentometric:≥94.0 to ≤106.0 %
- Water:≤2.5 %
- NMR (1H-NMR):Consistent with structure