DL-Arginine hydrochloride, anhydrous 98%
Catalog # ACRO446020050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Assay:97.5 to 102.5 %
- Loss on drying:≤3 %
Specification Test Results
About this item
:
:210.66 g/mol
Melting Pt:200…203 °C
Storage Temperature:Ambient
CAS Number:32042-43-6
:MFCD00063117
Formula:C₆H₁₄N₄O₂.HCl
MDL Number:MFCD00064549