Order Entry
Germany
ContactUsLinkComponent
Hydrogen chloride 3 M in CPME (cyclopentyl methyl ether), AcroSeal®
Flammable
Corrosive
Irritant
Hydrogen chloride 3 M in CPME (cyclopentyl methyl ether), AcroSeal®
Catalog # 445851000.
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Hydrogen chloride 3 M in CPME (cyclopentyl methyl ether), AcroSeal®
Catalog # 445851000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to light yellow Solution
  • Strength:
    2.85 to 3.30 M

Specification Test Results

About this item

 :
 :36.46 g/mol
Boiling Pt:106 °C
Density:0.895 g/cm³
Flash Pt:–1 °C
Storage Temperature:Ambient
UN:2920
ADR:8,I
 :GHS02
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324