2-Ethoxyethanol for analysis
:110-80-5
:MFCD00008590
:C₂H₅OCH₂CH₂OH
:MFCD00002869
:90.12 g/mol
:135 °C (760 mmHg)
:–70 °C
:0.93 g/cm³
:40 °C
:1171
:3,III
:GHS06
:GHS08
:GHS02
:Ambient
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.1 %
- GC:≥98.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Acidity/alkalinity:≤0.001 meq/g
- Calcium (Ca):≤1 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.5 ppm
- Lead (Pb):≤0.05 ppm
- Magnesium (Mg):≤0.2 ppm
- Potassium (K):≤0.5 ppm
- Silicon (Si):≤0.05 ppm
- Sodium (Na):≤2 ppm
- Sulfur (S):≤2 ppm
- Zinc (Zn):≤1 ppm