
Diclofenac sodium salt 98%
ACRO445250100
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- HPLC:≥97.5 %
- pH:6.5 to 7.5
:
:318.1 g/mol
:283…285 °C
:Ambient
:2811
:6.1,III
:GHS06
:15307-79-6
:C₁₄H₁₀Cl₂NNaO₂
:MFCD00082251