
3-Hydroxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 97%
Catalog # ACRO445050010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light yellow Solid
- Infrared spectrum:Conforms
- Melting point:84 to 90 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:278.11 g/mol
Melting Pt:85…87 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1073371-99-9
Formula:C₁₄H₁₉BO₅
MDL Number:MFCD08458200