
2-Nitrophenylboronic acid pinacol ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:190788-59-1
:
Formula:C₁₂H₁₆BNO₄
MDL Number:MFCD02179447
:249.07 g/mol
Melting Pt:45…47 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to brown Solid
- Infrared spectrum:Conforms
- Melting point:43 to 50 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure