4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:330794-35-9
:
Formula:C₁₈H₂₈BNO₄
MDL Number:MFCD06409942
:333.235 g/mol
Melting Pt:68…69 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white to off-white Solid
- Infrared spectrum:Conforms
- Melting point:66 to 71 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure