Order Entry
Germany
ContactUsLinkComponent
 
Hydrogen peroxide 35% (w/w) in water stabilised, Technical Grade
Catalog # ACRO444380050
CAS Number:  
undefined
Hydrogen peroxide 35% (w/w) in water stabilised, Technical Grade
Catalog # ACRO444380050
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Additional info:
    Stabilized
  • Titration with KMnO4:
    33 to 37 %

Specification Test Results

About this item

 :
Boiling Pt:108 °C (760 mmHg)
Melting Pt:–33 °C
Density:1.135 g/cm³
ADR:5.1,II
 :34.01 g/mol
UN:2014
 :GHS05
 :GHS07
 :GHS03
Storage Temperature:Refrigerator
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333