
Citric acid monohydrate, crystalline, Technical Grade
ACRO444370050
:
- Pack type:Plastic drum
- Pk:5 kg
- Seq:4114
- Appearance:White to almost white Crystalline powder, crystals or granules
- Water:7.5 to 9 %
- Titration with NaOH:99 to 101 %
:
:210.14 g/mol
:135…152 °C
:1,552 g/cm³ (20 °C)
:174 °C
:GHS07
:Ambient
:5949-29-1
:C₆H₈O₇.H₂O
:MFCD00149972