Order Entry
Germany
ContactUsLinkComponent
 
 
2-(Dimethylamino)thiazole-5-carboxaldehyde 97%
 
undefined
 
2-(Dimethylamino)thiazole-5-carboxaldehyde 97%

 

  • Appearance:
    Yellow to brown Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    72 to 78 °C
  • GC:
    ≥96.0 %