Order Entry
Germany
ContactUsLinkComponent
Methyl-2-pyrazinecarboxylate 97%
Irritant
Methyl-2-pyrazinecarboxylate 97%
Certificates
undefined
Irritant
Methyl-2-pyrazinecarboxylate 97%

Specification Test Results

  • Appearance:
    White to yellow to light orange Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    57 to 63 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together