ContactUsLinkComponent
L-Alanine-tert-butyl ester hydrochloride 99%
L-Alanine-tert-butyl ester hydrochloride 99%
Certificates
undefined
L-Alanine-tert-butyl ester hydrochloride 99%
Specification Test Results
  • Appearance:
    White to off-white Crystalline powder or solid
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥98.5 to ≤101.5 %
  • Specific optical rotation:
    +1.2° to +1.6° (20°C, 589 nm) (c=2 in EtOH)