0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Methyl-4-aminobutanoate hydrochloride
Methyl-4-aminobutanoate hydrochloride 99%
Methyl-4-aminobutanoate hydrochloride 99%
:
Thermo Fisher Scientific
Methyl-4-aminobutanoate hydrochloride 99%
:
Thermo Fisher Scientific
:
13031-60-2
:
:
C₅H₁₁NO₂·HCl
:
MFCD00043270
:
153.61 g/mol
:
118…119 °C
:
GHS07
:
Ambient
Appearance:
White to off-white Powder
Infrared spectrum:
Conforms
Titration Argentometric:
98.5 to 101.5 %
ACRO44291