Boc-L-tyrosine methyl ester 97%
:4326-36-7
:
:C₁₅H₂₁NO₅
:MFCD00191181
:295.3 g/mol
:106…107 °C
:Ambient
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Specific optical rotation:+48° to +52° (20°C, 589 nm) (c=1 in CHCl3)