
2-Phenylethylamine hydrochloride 98%
ACRO441910250
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 to ≤102.5 %
- NMR (1H-NMR):Consistent with structure
:
:157.64 g/mol
:220…223 °C
:GHS07
:Ambient
:156-28-5
:C₈H₁₁N
:MFCD00012893