ContactUsLinkComponent
N-Boc-cis-4-hydroxy-L-proline 97%
Irritant
Environment
N-Boc-cis-4-hydroxy-L-proline 97%
Certificates
undefined
Irritant
Environment
N-Boc-cis-4-hydroxy-L-proline 97%
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Specific optical rotation:
    -47° to -53° (20°C, 589 nm) (c=0.67 in MeOH)
  • Titration with NaOH:
    96 to 104 %