
COMU 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1075198-30-9
:
Formula:C₁₂H₁₉F₆N₄O₄P
MDL Number:MFCD11975052
:428.272 g/mol
Melting Pt:151.4…152.9 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:≥145.0 °C (Dec)
- Water:≤3.00 %
- NMR (1H-NMR):Consistent with structure