L(+)-Sodium lactate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:867-56-1
:
Formula:C₃H₅NaO₃
MDL Number:MFCD00066576
:112.06 g/mol
Melting Pt:161.6…165 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder or solid
- Infrared spectrum:Conforms
- Melting point:160 to 166 °C
- Water:≤1.0 %
- Specific optical rotation:-12° to -13° (20 °C, 589 nm) (C=1 in H2O)
- Titration with HClO4:≥98.5 %