Order Entry
Germany
ContactUsLinkComponent
2-Amino-2-thiazoline 97%
2-Amino-2-thiazoline 97%
Certificates
undefined
2-Amino-2-thiazoline 97%

Specification Test Results

  • Appearance:
    White to off-white to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    78 to 83 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Related Information