Order Entry
Germany
ContactUsLinkComponent
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 97%
Corrosive
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 97%
Certificates
undefined
Corrosive
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 97%

Specification Test Results

  • Appearance:
    Colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4600 to 1.4630 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information