Boc-L-alaninal 98%
Catalog # ACRO438470010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Specific optical rotation:-29° to -39° (20°C, 589 nm) (C=2 in methanol)
Specification Test Results
About this item
:
:173.212 g/mol
Melting Pt:86.4…87.6 °C
Storage Temperature:Freezer
CAS Number:79069-50-4
Formula:C₈H₁₅NO₃
MDL Number:MFCD00143786