(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122536-76-9
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD00143194
:186.26 g/mol
Melting Pt:77.8…80.7 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:72 to 82 °C
- GC:≥98.5 %
- Specific optical rotation:-17° to -23° (20 °C, 589 nm) (c=1, ethanol)