(±)-3-(Boc-amino)piperidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:172603-05-3
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD03839941
:200.28 g/mol
Melting Pt:104.6…112.6 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- GC:≥96.0 %