
5-Bromo-2-methoxypyridine-3-carboxaldehyde 97%
Catalog # ACRO437390050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to brown Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
About this item
:
:216.034 g/mol
Melting Pt:102…104 °C
:GHS07
Storage Temperature:Ambient
CAS Number:103058-87-3
Formula:C₇H₆BrNO₂
MDL Number:MFCD05663521