Aluminium 99.9995% (trace metals basis), shot
Catalog # ACRO437070250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Silver grey Shots
- Total trace metal impurities:≤5 ppm
Specification Test Results
About this item
:
:26.98 g/mol
Boiling Pt:2327 °C (760 mmHg)
Melting Pt:660 °C
Density:2.7 g/cm³
Storage Temperature:Ambient
CAS Number:7429-90-5
Formula:Al
MDL Number:MFCD00134029