Nickel 99.995% (trace metals basis), powder
Catalog # ACRO436780500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Appearance:Black Powder
- Total trace metal impurities:≤50 ppm
Specification Test Results
About this item
:
:58.69 g/mol
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1455 °C
Density:8,9 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3089
ADR:4.1,II
:GHS02
:GHS07
:GHS08
CAS Number:7440-02-0
Formula:Ni
MDL Number:MFCD00011137