
(S)-1-Boc-3-hydroxypiperidine 97%
Catalog # ACRO436440050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to tan Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Specific optical rotation:+9° to +11° (20°C, 589 nm) (C=1 in chloroform) / +21° to +24° (20°C, 589 nm) (c=5 in MeOH)
Specification Test Results
About this item
:
:201.27 g/mol
Melting Pt:48.9…51.7 °C
Flash Pt:110 °C
:GHS07
Storage Temperature:Ambient
CAS Number:143900-44-1
Formula:C₁₀H₁₉NO₃
MDL Number:MFCD04115307