Methyl (R)-(-)-3-hydroxyisobutyrate 98%
Catalog # ACRO436180050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Specific optical rotation:-23° to -29° (20°C, 589 nm) (c=4, methanol)
Specification Test Results
About this item
:
:118.1311 g/mol
Boiling Pt:60 °C (4Torr)
Density:1.07 g/cm³
Flash Pt:81 °C
Storage Temperature:Ambient
CAS Number:72657-23-9
Formula:C₅H₁₀O₃
MDL Number:MFCD00063450